Order Entry
Belgium
ContactUsLinkComponent
n-Heptane ≥99%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Flammable
Health
Irritant
Environment
n-Heptane ≥99%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Health
Irritant
Environment
n-Heptane ≥99%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 5
  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤10
  • ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):
    ≤5
  • Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water):
    ≥99.0 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤1.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤100 ppm

Specification Test Results

Related Information

Frequently Bought Together