Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
 
n-Heptane ≥99.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
  9177-68
 :  Avantor
 :  
undefined
 
 
 
 
n-Heptane ≥99.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
  9177-68
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤0.40
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤197
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.2 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Assay (by GC, corrected for water):
    ≥99.0 %
  • Residue after Evaporation:
    ≤2.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤0.01 %

 

 

 :
 :100.20 g/mol
 :13,04679
 :98.4 °C (1013 hPa)
 :–90.6 °C
 :0.6838 g/cm³ (20 °C)
 :–4 °C
 :Ambient
 :205-563-8
 :1206
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :142-82-5
 :H₃C(CH₂)₅CH₃
 :MFCD00009544