n-Heptane, anhydrous 99+%
:142-82-5
:
:H₃C(CH₂)₅CH₃
:MFCD00009544
:100.20 g/mol
:14,4659
:97…99 °C
:–91 °C
:0.684
:Ambient
:205-563-8
:1206
:3,II
:Packaged under argon in resealable ChemSeal™ bottles.
:–4 °C (24 °F)
:GHS02
:GHS08
:GHS07
:GHS09
Frequently Bought Together