n-Heptane 99+% for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
:100.20 g/mol
Merck Index:14,4659
Boiling Pt:97…99 °C
Melting Pt:–91 °C
Density:0.684
Flash Pt:–4 °C (24 °F)
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Residual water:0.03%
- Residue on Evaporation:3ppm
- n-Heptane:96% (info)
Specification Test Results
Related Information
Frequently Bought Together