n-Heptane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–91 °C
Density:0.683 g/cm³
Flash Pt:–4 °C
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 %
- GC:≥99.0 %
- UV:at 200 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
Specification Test Results
Frequently Bought Together