

n-Decane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-18-5
:
Formula:H₃C(CH₂)₈CH₃
MDL Number:MFCD00008954
:142.28 g/mol
Boiling Pt:174.1 °C (1013 hPa)
Melting Pt:–30 °C
Density:0.73 g/cm³ (20 °C)
Flash Pt:46 °C
UN:2247
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:204-686-4
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.00005 g/l (25 °C)
- Melting Point:-30 °C
- Molar Mass:142.28 g/mol
- Boiling Point:174 °C
- Vapor Pressure:1 hPa (16 °C)
- Flash Point:51 °C
- Explosion Limit:0.7 - 5.4 %(V)
- Density:0.73 g/cm3 (20 °C)
- Ignition Point:208 °C
- Viscosity Kinematic:- 7 mm2/s (40 °C)
Specification Test Results
Related Information
Frequently Bought Together