Sodium dodecyl sulphate (SDS)
SERA20770.01
: Serva
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:288.38 g/mol
:14,08636
:204…207 °C
:1.1 g/cm³ (20 °C)
:>100 °C
:Ambient
:205-788-1
:1325
:4.1,II
:GHS02
:GHS06
:151-21-3
:CH₃(CH₂)₁₁OSO₃Na
:MFCD00036175