Sodium dodecyl sulphate (SDS)
500 g
Catalog # DG093
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
- CAS Number:151-21-3
- Formula:CH₃(CH₂)₁₁OSO₃Na
- MDL Number:MFCD00036175
- Molecular Weight:288.38 g/mol
- Merck Index:14,08636
- Melting Pt:204…207 °C
- Density:1.1 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:205-788-1
- UN:1325
- ADR:4.1,II
Specifications
Specifications
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



