Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥99.0% for biochemistry, specially purified
Flammable
Irritant
Corrosive
Sodium dodecyl sulphate (SDS) ≥99.0% for biochemistry, specially purified
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
Corrosive
Sodium dodecyl sulphate (SDS) ≥99.0% for biochemistry, specially purified
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.0 %
  • Assay (C12 compounds)(calc.on anhydrous):
    Min. 98.5 %
  • IR Spectrum:
    Passes test
  • Cl (Chloride):
    Max. 0.03 %
  • PO4 (Phosphate):
    Max. 1 ppm
  • Pb (Lead):
    Max. 2 ppm
  • Absorbance (220-350 nm) (3 %):
    Max. 0.1

Specification Test Results

Related Information

Frequently Bought Together