1-Octadecanethiol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2885-00-9
:
Formula:C₁₈H₃₈S
MDL Number:MFCD00004886
:286.57 g/mol
Boiling Pt:204 °C (1013 hPa)
Melting Pt:28…31 °C
Density:0.8475 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:220-744-1
ADR:9,
:M-Clarity™ quality level = MQ200
UN:3335
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:28 - 31 °C
- Molar Mass:286.55 g/mol
- Boiling Point:366 °C
- Vapor Pressure:- 0.5 hPa (20 °C)
- Flash Point:185 °C
- Refractive Index:1.4645 (20 °C, 589 nm)
- Density:0.847 g/cm3 (20 °C)
- Ignition Point:200 °C
Specification Test Results
Related Information
Frequently Bought Together