Order Entry
Belgium
ContactUsLinkComponent
Ethylene glycol ≥99.0% Reag. Ph. Eur.
Irritant
Ethylene glycol ≥99.0% Reag. Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Ethylene glycol ≥99.0% Reag. Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Appearance:
    Clear viscous liquid
  • Relative density:
    1.113 - 1.115
  • Refractive index (20°C) ( ± 1.432):
    Passes test
  • Boiling point (± 198 °C):
    Passes test
  • Acidity:
    Passes test
  • Water:
    Max. 0.2 %

Specification Test Results

Frequently Bought Together