Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methanol

Methanol

Flammable
Toxic
Health
Marketsource
Supplier:   Roth Carl
Methanol
Supplier:  Roth Carl
Marketsource
Certificates
Methanol

6 Options Available Below

Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
    Marketsource
    Marketsource
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L

    Specifications:
    Marketsource
    Marketsource
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.