Methanol ≥99.8%
Glass bottle, 1 L
Supplier: VWR Chemicals
Methanol ≥99.8%
Catalog # 20834.291
Supplier: VWR Chemicals
CAS Number: 67-56-1
Jump to:
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0003 meq/g
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.05 %
- Absorbance (220 nm):Max. 0.4
- Absorbance (240 nm):Max. 0.15
- Absorbance (260 nm):Max. 0.02
- Transmittance (220 nm):Min. 45 %
- Transmittance (240 nm):Min. 75 %
- Transmittance (260 nm):Min. 96 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:20834.291
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



