Order Entry
Belgium
ContactUsLinkComponent
Methanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Flammable
Toxic
Health
Methanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Toxic
Health
Methanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay:
    Min. 99,8 %
  • Acidity:
    Max. 0,00007 meq/g
  • Alkalinity:
    Max. 0,0001 meq/g
  • Density (20/4):
    0,7900 --> 0,7920
  • Non-volatile residue:
    Max. 5 ppm
  • Water:
    Max. 0,0500 %
  • Transmittance (210 nm):
    Min. 60,0 %
  • Transmittance (220 nm):
    Min. 80,0 %
  • Transmittance (230 nm):
    Min. 90,0 %
  • Transmittance (235 nm):
    Min. 92,0 %
  • Transmittance (240 nm):
    Min. 95,0 %
  • Transmittance (250 nm):
    Min. 98,0 %
  • Transmittance (260 nm):
    Min. 98,0 %

Specification Test Results

Frequently Bought Together