Order Entry
Belgium
ContactUsLinkComponent
Methanol, anhydrous ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS for Karl Fischer titration, J.T.Baker®
Flammable
Toxic
Health
Methanol, anhydrous ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS for Karl Fischer titration, J.T.Baker®
Catalog # 9049-02
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol, anhydrous ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS for Karl Fischer titration, J.T.Baker®
Catalog # 9049-02
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 99.8 %
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • ACS - Assay (CH₃OH) (by GC, corrected for water):
    ≥99.8 %
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Solubility in H₂O:
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Water (H₂O):
    ≤0.007 %
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • Carbonyl Compounds - Acetaldehyde:
    ≤0.001 %
  • Carbonyl Compounds - Acetone:
    ≤0.001 %
  • Carbonyl Compounds - Formaldehyde:
    ≤0.001 %
  • Titrable Acid (µeq/g):
    ≤0.3
  • Titrable Base (µeq/g):
    ≤0.2

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595