Order Entry
Belgium
ContactUsLinkComponent
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Irritant
Flammable
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Catalog # 41322-1L
CAS Number:  
Irritant
Flammable
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Catalog # 41322-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Vapor Density:
    1.59 (vs air)
  • Appearance:
    complying
  • Appearance of the solution:
    complying
  • Assay:
    Min. 99.5 Vol.%
  • Density (D 20/20):
    0.790 - 0.793
  • Identity (IR):
    complying
  • Acidity or alkalinity:
    complying
  • Non-volatile matter:
    Max. 25 ppm
  • Residual solvents:
    complying
  • Volatile impurities (GC):
    complying
  • Acetaldehyde & Acetale (GC):
    Max. 10 ppm
  • Benzene (GC):
    Max. 2 ppm
  • Methanol (GC):
    Max. 200 ppm
  • Sum all contaminations (GC):
    Max. 300 ppm
  • Absorbance at 240 nm:
    Max. 0.40
  • Absorbance 250-260 nm:
    Max. 0.30
  • Absorbance 270-340 nm:
    Max. 0.10

Specification Test Results

About this item

 :
 :46.07 g/mol
Merck Index:12,03806
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.790…0.791 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
 :GHS07
 :GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568