Order Entry
Belgium
ContactUsLinkComponent
Ethanol (untaxed) ~96% (v/v), CHROMASOLV™ ACS for UV spectrophotometry extra fine spirit, E15 o 1, Riedel-de Haën™
Ethanol (untaxed) ~96% (v/v), CHROMASOLV™ ACS for UV spectrophotometry extra fine spirit, E15 o 1, Riedel-de Haën™
   Sustainable Options Available
Certificates
Ethanol (untaxed) ~96% (v/v), CHROMASOLV™ ACS for UV spectrophotometry extra fine spirit, E15 o 1, Riedel-de Haën™
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Purity without addives GC):
    Min. 99.8 %
  • Refractive index (n 20/D):
    1.362 - 1.364
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    5.0 - 7.0 %
  • Titrable Acid:
    Max. 0.0005 meq/g
  • Titrable Base:
    Max. 0.0002 meq/g
  • Absorbance at 270 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.03
  • Absorbance at 230 nm:
    Max. 0.1
  • Absorbance at 220 nm:
    Max. 0.25
  • Absorbance at 210 nm:
    Max. 0.5
  • KMnO₄ reducing matter:
    complying
  • Acetone (GC):
    Max. 10 ppm
  • Methanol (GC):
    Max. 0.1 %
  • 2-propanol (GC):
    Max. 30 ppm
  • Reaction against H₂SO₄:
    complying
  • Mixable with H₂O:
    complying
  • APHA:
    Max. 10

Specification Test Results

Related Information

Frequently Bought Together