

Ethanol absolute, Uvasol® for spectroscopy, Supelco®
: Merck
:64-17-5
:
:C₂H₅OH
:MFCD00003568
:46.07 g/mol
:12,03806
:78.3 °C (1013 hPa)
:–117 °C
:0.7895 g/cm³ (20 °C)
:200-578-6
:1170
:3,II
:M-Clarity™ quality level = MQ100
:12 °C
:01-2119457610-43
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Solubility:(20 °C) completely miscible
- Melting Point:-114.5 °C
- Molar Mass:46.07 g/mol
- Boiling Point:78.3 °C (1013 hPa)
- Vapor Pressure:59 hPa (20 °C)
- Flash Point:12 °C
- Refractive Index:1.36
- Explosion Limit:3.5 - 15%(V)
- Density:0.790 - 0.793 g/cm³ (20 °C)
- pH:7.0 (10 g/L, H₂O, 20 °C)
- Ignition Point:363 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together