Order Entry
Belgium
ContactUsLinkComponent
2-Propanol ≥99.0%, GPR RECTAPUR®
Flammable
Irritant
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Acidity:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Benzene and related substances:
    Passes test
  • IR Spectrum:
    Passes test
  • Peroxides:
    Passes test
  • Boiling point:
    82.2 - 82.6 °C
  • Density (20/20):
    0.785 - 0.789
  • Density (25/25):
    0.783 - 0.790
  • Refractive index (20°C):
    1.376 - 1.378
  • Evaporation residue:
    Max. 20 ppm
  • Water:
    Max. 0.5 %

Specification Test Results

Frequently Bought Together