

2-Propanol ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., USP, Riedel-de Haën™
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:13,05228
:77…83 °C (1.013 hPa)
:–90 °C
:0.784…0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Appearance:complying
- Identity (IR):complying
- Assay (GC):Min. 99.5 %
- Density (D 20/20):0.785 - 0.788
- Density (D 25/25):0.783 - 0.787
- Refractive index (n 20/D):1.377 - 1.378
- Non-volatile matter:Max. 20 ppm
- Water (Karl Fischer):Max. 0.2 %
- Acidity or alkalinity:complying
- Acidity:complying
- Iron (Fe):Max. 0.0001 %
- Heavy metals (as Pb):Max. 0.0002 %
- Aldehydes, Ketones (as CH₃CocH₃):Max. 0.05 %
- Benzene:Max. 0.0001 %
- Benzene and related substances:complying
- Methanol (GC):Max. 0.02 %
- Peroxides:complying
- Absorbance:complying
- Volatile impurities (GC):complying
- Residual solvents:complying
- Autoignition Temperature:750 °F
Frequently Bought Together