

2-Propanol ≥99.7% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9334-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 5
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- Assay (CH₃CHOHCH₃) (by GC, corrected for water):≥99.7 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Water (by KF, coulometric):≤0.1 %
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:Packaged under inert gas.
REACH:01-2119457558-25
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674