

2-Propanol 99+% for spectroscopy
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:13,05228
:81…83 °C (760 mmHg)
:–89.5 °C
:0.785 g/cm³
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.0 %
- UV:at 205 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0003 %
Frequently Bought Together