Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

1-Tetradecanol

Marketsource
Supplier:   Roth Carl
1-Tetradecanol
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
  • CAS Number:112-72-1
  • Formula:H₃C(CH₂)₁₂CH₂OH
  • MDL Number:MFCD00004757
  • Molecular Weight:214.39 g/mol
  • Merck Index:13,06361
  • Boiling Pt:289 °C (1013 hPa)
  • Melting Pt:37…39 °C
  • Density:0.8 g/cm³ (60 °C)
  • Storage Temperature:Ambient
  • EINECS:204-000-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    Marketsource
    Marketsource