Order Entry
Belgium
ContactUsLinkComponent
1-Propanol ≥99%, GPR RECTAPUR®
1-Propanol ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
1-Propanol ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Acidity:
    Max. 0.0005 meq/g
  • Boiling point:
    96.9 - 97.5 °C
  • Density (20/4):
    0.801 - 0.804
  • Substances coloured by H2SO4:
    Max. 150 APHA
  • Evaporation residue:
    Max. 100 ppm
  • Methanol:
    Max. 0.2 %
  • Water:
    Max. 0.2 %

Specification Test Results

Frequently Bought Together