Order Entry
Belgium
ContactUsLinkComponent
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 99 %
  • Boiling Range:
    max. 2.0 °C
  • Color (APHA):
    max. 10
  • Density (g/ml) at 25°C:
    0.800 - 0.804
  • Ethanol (C2H5OH) (by GC):
    max. 0.05 %
  • Free Acid (as C2H5COOH):
    max. 0.001 %
  • Methanol (by GC) (CH3OH):
    max. 0.05 %
  • Recorded Boiling Point:
    97.2 °C
  • Residue after Evaporation:
    max. 0.005 %
  • Substances Darkened by H2SO4:
    passes test
  • Substances Reducing KMnO4 (as O):
    max. 0.0003 %
  • Water (H2O):
    max. 0.1 %
  • 2-Propanol (C3H7OH) (by GC):
    max. 0.05 %
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together