Order Entry
Belgium
ContactUsLinkComponent
1-Propanol 99% for spectrophotometry
1-Propanol 99% for spectrophotometry
Catalog # 22933.K2
CAS Number:  
undefined
1-Propanol 99% for spectrophotometry
Catalog # 22933.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

CAS Number:71-23-8
 :
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941
 :60.1 g/mol
Merck Index:14,7842
Boiling Pt:97 °C
Melting Pt:–127 °C
Density:0.804
Flash Pt:15 °C (59 °F)
Storage Temperature:Ambient
EINECS:200-746-9
UN:1274
ADR:3,II
 :GHS02
 :GHS05
 :GHS07