1-Hexanol ≥98% for synthesis, Sigma-Aldrich®
: Merck
:111-27-3
:
:H₃C(CH₂)₄CH₂OH
:MFCD00002982
:102.18 g/mol
:13,04715
:156 °C (1013 hPa)
:–45 °C
:0.8322 g/cm³ (20 °C)
:203-852-3
:2282
:3,III
:M-Clarity™ quality level = MQ200
:60 °C
:GHS02
:GHS07
:Ambient
- Assay (GC, area%):≥ 98 %
- Density (d 20 °C/ 4 °C):0.818 - 0.819
- Identity (IR):passes test
- Synonyms:1-Hexanol
- Chemical Formula:CH3(CH2)5OH
Frequently Bought Together