Order Entry
Belgium
ContactUsLinkComponent
 
Flammable
Irritant
Corrosive
1-Butanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83633.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Corrosive
1-Butanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83633.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Acidity (as CH3COOH):
    Max. 0.002 %
  • Colour:
    Max. 10 APHA
  • Residue after evaporation:
    Max. 0.0005 %(m)
  • Water (KF):
    Max. 0.05 %(m)
  • Transmittance (210 nm):
    Min. 20 %
  • Transmittance (230 nm):
    Min. 70 %
  • Transmittance (270 nm):
    Min. 90 %
  • Transmittance (310 nm):
    Min. 98 %

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119484630-38
 :GHS02
 :GHS07
 :GHS05
CAS Number:71-36-3
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964