


1-Butanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:14,1540
Boiling Pt:117…118 °C
Melting Pt:–90…–89 °C
Density:0.810
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
:Packaged under argon in resealable ChemSeal™ bottles.
Flash Pt:35 °C (95 °F)
REACH:01-2119484630-38
:GHS05
:GHS02
:GHS07
Frequently Bought Together