Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Flammable
Irritant
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Catalog # 83671.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Catalog # 83671.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle with septum cap
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Residue on evaporation:
    Max. 0.0003 %
  • Water:
    Max. 0.0050 %

Specification Test Results

About this item

CAS Number:60-29-7
 :
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,I
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–40 °C
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 7-10 ppm
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
 :GHS02
 :GHS07