Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether ≥99.5% stabilised, PESTINORM® for pesticide residue analysis
Flammable
Irritant
Diethyl ether ≥99.5% stabilised, PESTINORM® for pesticide residue analysis
Catalog # 83659.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether ≥99.5% stabilised, PESTINORM® for pesticide residue analysis
Catalog # 83659.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.50 %
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.0200 %
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
ADR:3,I
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :Stabilised with ethanol < 1%
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646