Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether ≥99.5% stabilised, HiPerSolv CHROMANORM® for HPLC
Flammable
Irritant
Diethyl ether ≥99.5% stabilised, HiPerSolv CHROMANORM® for HPLC
Catalog # 83624.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether ≥99.5% stabilised, HiPerSolv CHROMANORM® for HPLC
Catalog # 83624.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.50 %
  • Water:
    Max. 0.050 %
  • Evaporation residue:
    Max. 0.0005 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Peroxides:
    Max. 5 ppm
  • Colour number (Hazen):
    Max. 10
  • Transmittance (220 nm):
    Min. 30.0 %
  • Transmittance (230 nm):
    Min. 50.0 %
  • Transmittance (250 nm):
    Min. 80.0 %
  • Transmittance (270 nm):
    Min. 90.0 %
  • Transmittance (300 nm):
    Min. 98.0 %

Specification Test Results

About this item

CAS Number:60-29-7
 :
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,I
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–40 °C
 :Stabilised with ethanol 1 %
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
 :GHS02
 :GHS07