Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether ≥99% stabilised, GPR RECTAPUR® peroxide-free
Diethyl ether ≥99% stabilised, GPR RECTAPUR® peroxide-free
Catalog # 23806.328
Supplier:  VWR Chemicals
CAS Number:  
undefined
Diethyl ether ≥99% stabilised, GPR RECTAPUR® peroxide-free
Catalog # 23806.328
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Assay (Ionol/BHT):
    4 - 10 ppm
  • Acidity or alkalinity:
    Max. 0.0002 meq/g
  • Aldehydes + ketones (as HCHO):
    Max. 5 ppm
  • Evaporation residue:
    Max. 20 ppm
  • Peroxides (as H2O2):
    Max. 0.15 ppm
  • Water:
    Max. 0.2 %

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 4-7 ppm
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646