Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Flammable
Irritant
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Catalog # 1.00930.1000
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Catalog # 1.00930.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    69 g/L (20 °C)
  • Melting Point:
    -116.3 °C
  • Molar Mass:
    74.12 g/mol
  • Boiling Point:
    34.6 °C (1013 hPa)
  • Vapor Pressure:
    587 hPa (20 °C)
  • Flash Point:
    -40 °C
  • Explosion Limit:
    1.7 - 36%(V)
  • Density:
    0.71 g/cm³
  • Ignition Point:
    180 °C
  • Saturation Concentration:
    1776 g/m³ (20 °C) Air
  • Evaporation Number:
    1
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
 :M-Clarity™ quality level = MQ100
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646