Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, ultra-low water content ≥99.0% (by GC) unstabilised, BakerDRY™, J.T.Baker®
Diethyl ether, ultra-low water content ≥99.0% (by GC) unstabilised, BakerDRY™, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Diethyl ether, ultra-low water content ≥99.0% (by GC) unstabilised, BakerDRY™, J.T.Baker®
Supplier:  Avantor

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay ((C₂H₅)₂O) (by GC):
    ≥99.0 %
  • ACS - Alcohol (C₂H₅OH):
    ≤0.05 %
  • ACS - Carbonyl Compounds (as HCHO) (by polarography):
    ≤0.001 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Peroxide (as H₂O₂):
    ≤1 ppm
  • ACS - Residue after Evaporation:
    ≤5 ppm
  • ACS - Titrable Acid (µeq/g):
    ≤0.2
  • ACS - Water (by KF, coulometric):
    ≤10.0 ppm

Specification Test Results

Frequently Bought Together