Order Entry
Belgium
ContactUsLinkComponent
Trifluoroacetic acid 100%, HiPerSolv CHROMANORM® for HPLC
Trifluoroacetic acid 100%, HiPerSolv CHROMANORM® for HPLC
Catalog # 153112E
Supplier:  VWR Chemicals
CAS Number:  
undefined
Trifluoroacetic acid 100%, HiPerSolv CHROMANORM® for HPLC
Catalog # 153112E
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    Min. 99.80 %
  • Colour value:
    Max. 10 APHA
  • Non-volatile matter:
    Max. 0.005 %
  • Transmittance (210 nm):
    Min. 30 %
  • Transmittance (220 nm):
    Min. 60 %
  • Transmittance (230 nm):
    Min. 85 %
  • Transmittance (240 nm):
    Min. 95 %
  • Transmittance (250 nm):
    Min. 99 %

Specification Test Results

About this item

 :
 :114.02 g/mol
Merck Index:13,09754
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
 :Packaged under nitrogen.
 :GHS07
 :GHS05
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169