

Trifluoroacetic acid ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-05-1
:
Formula:C₂HF₃O₂
MDL Number:MFCD00004169
:114.02 g/mol
Merck Index:14,9681
Boiling Pt:72…73 °C
Melting Pt:–15 °C
Density:1.480
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07
Related Information
Frequently Bought Together