Order Entry
Belgium
ContactUsLinkComponent
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    99.0 - 99.6 %
  • Boiling point:
    59.5 - 60.5 °C
  • Density (20/4):
    1.473 - 1.489
  • Ethanol:
    0.1 - 1.0 %
  • Evaporation residue:
    Max. 50 ppm
  • Free chlorine:
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Water:
    Max. 100 ppm

Specification Test Results

Frequently Bought Together