Chloroform, LiChrosolv® for HPLC
Supplier: Merck
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:200-663-8
REACH:05-2114614906-45
:GHS06
:GHS08
Related Information
Frequently Bought Together