Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
About this item
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39.8 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1.322 g/cm³ (20 °C)
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with ethanol 0.1 %
- Storage Temperature:Ambient
- UN:1593
- REACH:01-2119480404-41
6 Options Available Below
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39.8 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1.322 g/cm³ (20 °C)
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with ethanol 0.1 %
- Storage Temperature:Ambient
- UN:1593
- REACH:01-2119480404-41
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 216L Metal drum for liquids UN X (ZPACK10843202) 28L Metal drum for liquids UN X (ZPACK12124128) 30L Metal drum for liquids UN X (ZPACK12124125)
More
Specifications for Product Family
Specification Test Results
- Assay:Min. 99 %
- Acidity:Max. 0.0004 meq/g
- Boiling point:39.0 - 40.5 °C
- Density (20/4):1.322 - 1.327
- Evaporation residue:Max. 20 ppm
- Water:Max. 0.02 %



