Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Methylene chloride
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Stabilised with ethanol 0.15 %
:84.93 g/mol
Storage Temperature:Ambient
REACH:01-2119480404-41
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 28L Metal drum for liquids UN X (ZPACK12124128) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
spec_VWRC25629.295 spec_VWRC25629.295 spec_VWRC25629.364 spec_VWRC25629.364 spec_VWRC25629.466
Product specification
More
Frequently Bought Together