Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Health
Irritant
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Irritant
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Boiling point:
    39.0 - 40.5 °C
  • Acidity:
    Max. 0.0004 meq/g
  • Density (20/4):
    1.322 - 1.327
  • Evaporation residue:
    Max. 10 ppm
  • Free chlorine:
    Max. 0.3 ppm
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Water:
    Max. 0.02 %
  • Cl (Chloride):
    Max. 3 ppm

Specification Test Results

Frequently Bought Together