Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Dichloromethane ≥99.8% stabilised, Suprasolv® hypergrade for GC-ECD/FID, Supelco®
 :  Merck
 
undefined
 
 
Dichloromethane ≥99.8% stabilised, Suprasolv® hypergrade for GC-ECD/FID, Supelco®
 :  Merck
 

 

  • Purity (GC):
    ≥99.9%
  • Identity (IR):
    conforms
  • Evaporation Residue:
    ≤3.0 mg/L
  • Water:
    ≤0.005%
  • Colour:
    ≤10 Hazen
  • GC/ECD (Retention Range 1,2,4-Tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)):
    ≤2 pg/ml
  • GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)):
    ≤2 ng/ml
  • GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning Area 30 - 600 amu, Individual Signals (n-Tetradecane Standard)):
    ≤2 ng/ml

 

Frequently Bought Together