

Dichloromethane stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
:75-09-2
:
:CH₂Cl₂
:MFCD00000881
:84.93 g/mol
:12,06140
:39.8 °C (1013 hPa)
:–95 °C
:1.322 g/cm³ (20 °C)
:200-838-9
:1593
:6.1,III
:M-Clarity™ quality level = MQ300
:Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
:GHS08
:GHS07
:Ambient
- Solubility:20 g/l (20 °C)
- Melting Point:-95 °C
- Molar Mass:84.93 g/mol
- Boiling Point:40 °C (1013 hPa)
- Vapor Pressure:475 hPa (20 °C)
- Refractive Index:1.42
- Explosion Limit:13 - 22 %(V)
- Density:1.33 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:605 °C DIN 51794
- Evaporation number:1.9
Frequently Bought Together