Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 7305.1000
Supplier:  Avantor
CAS Number:  
undefined
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 7305.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99.5 %
  • Appearance:
    clear
  • Color (APHA):
    max. 10
  • Free Halogens:
    passes test
  • Preservative (as Amylene):
    about 20 ppm
  • Residue after Evaporation:
    max. 5 ppm
  • Titrable Acid (meq/g):
    max. 0.0003
  • Water (H2O):
    max. 0.02 %
  • at 235 nm:
    max. 1.00
  • at 240 nm:
    max. 0.35
  • at 250 nm:
    max. 0.10
  • at 260 nm:
    max. 0.04
  • at 340-400 nm:
    max. 0.01

Specification Test Results

About this item

Synonyms: Methylene chloride
 :
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :84.93 g/mol
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881