Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Methylene chloride
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
ADR:6.1,III
:84.93 g/mol
UN:1593
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (by GC) (corrected for water):min. 99.5 %
- Appearance:clear
- Color (APHA):max. 10
- Free Halogens:passes test
- Residue after Evaporation:max. 0.002 %
- Titrable Acid (meq/g):max. 0.0002
- Water (H2O):max. 0.02 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Specification Test Results
Frequently Bought Together