Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dichloroethane
1,2-Dichloroethane ≥99% for HPLC
1,2-Dichloroethane ≥99% for HPLC
:
Thermo Fisher Scientific
1,2-Dichloroethane ≥99% for HPLC
:
Thermo Fisher Scientific
:
107-06-2
:
:
ClH₂CCH₂Cl
:
MFCD00000963
:
98.96 g/mol
:
14,3797
:
83 °C
:
–35 °C
:
1.256
:
15 °C (59 °F)
:
Ambient
:
203-458-1
:
1184
:
3,II
:
GHS02
:
GHS08
:
GHS07
msds_ALFA022918
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Rota® Duo, pH indicator papers
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
: VWR Chemicals
n-Pentane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals