Order Entry
Belgium
ContactUsLinkComponent
Ammonium acetate ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Ammonium acetate ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Ammonium acetate ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 97 %
  • Insoluble Matter:
    max. 0.005 %
  • Nitrate (NO3):
    max. 0.001 %
  • pH of 5% Solution at 25°C:
    6.7 - 7.3
  • Residue after Ignition:
    max. 0.01 %
  • Sulfate (SO4):
    max. 0.001 %
  • Trace Impurities (in ppm):
    max. 5
  • Chloride (Cl):
    max. 5
  • Iron (Fe):
    max. 5
  • Heavy Metals (as Pb):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together