Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Potassium acetate

Marketsource
Supplier:   Serva
Potassium acetate
Supplier:  Serva
Marketsource
Certificates
Potassium acetate
Potassium acetate

2 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid potassium salt
  • CAS Number:127-08-2
  • Formula:CH₃COOK
  • MDL Number:MFCD00012458
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,07687
  • Melting Pt:304 °C
  • Density:1.57 g/cm³ (25 °C)
  • Flash Pt:>250 °C
  • Storage Temperature:Ambient
  • EINECS:204-822-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 76,00 €
    76,00 €
    1 * 500 g
       76,00 €

    Specifications:
    Marketsource
    Marketsource