Potassium acetate
25 kg
Catalog # LOHM107204.9025
Supplier: LOHMANN
Jump to:
Product Details & Documents
Synonyms: Acetic acid potassium salt
- CAS Number:127-08-2
- Formula:CH₃COOK
- MDL Number:MFCD00012458
- Molecular Weight:98.14 g/mol
- Merck Index:13,07687
- Melting Pt:304 °C
- Density:1.57 g/cm³ (25 °C)
- Flash Pt:>250 °C
- Storage Temperature:Ambient
- EINECS:204-822-2
Specifications
Specifications
- Pk:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



